missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate, 97%, Thermo Scientific Chemicals

Catalog Number p-7058324
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
22232-54-8
C7H10N2O2S
186.229
MFCD00027421
CFOYWRHIYXMDOT-UHFFFAOYSA-N
carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat
31072
CHEBI:617099
ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate
CCOC(=O)N1C=CN(C1=S)C
This item is not returnable. View return policy
22232-54-8
C7H10N2O2S
186.229
MFCD00027421
CFOYWRHIYXMDOT-UHFFFAOYSA-N
carbimazole, athyromazole, carbimazol, carbethoxymethimazole, neomercazole, carbinazole, neo-thyreostat, basolest, carbimazolum, thyrostat
31072
CHEBI:617099
ethyl 3-methyl-2-sulfanylideneimidazole-1-carboxylate
CCOC(=O)N1C=CN(C1=S)C

Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate is used as thyroid inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate is used as thyroid inhibitor.

Solubility
Soluble in water, ethanol, chloroform and acetone. Sightly soluble in water.

Notes
Light sensitive. Keep the container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY
Melting Point 122°C to 126°C
pH 5.0 to 7.0
Quantity 5 g
Beilstein 144339
Sensitivity Light sensitive
Merck Index 14,1798
Solubility Information Soluble in water,ethanol,chloroform and acetone. Sightly soluble in water.
Formula Weight 186.24
Percent Purity 97%
Chemical Name or Material Ethyl 3-methyl-2-thionoimidazoline-1-carboxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'