missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-chloro-5-pyridylglyoxylate, 97%, Thermo Scientific Chemicals

Catalog Number p-7033142
112.67 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
902837-55-2
C9H8ClNO3
213.617
MFCD07781188
HRFPVENOVYSKHE-UHFFFAOYSA-N
ethyl 6-chloro-3-pyridylglyoxylate, ethyl 2-6-chloropyridin-3-yl-2-oxoacetate, ethyl 2-chloro-5-pyridylglyoxylate, ethyl 6-chloropyridin-3-yl oxo acetate, ethyl6-chloro-3-pyridylglyoxylate, ethyl 6-chloro-3-pyridinyl oxoacetate, 6-chloro-3-pyridylglyoxylic acid ethyl ester, ethyl 2-6-chloro 3-pyridyl-2-oxoacetate
42553168
ethyl 2-(6-chloropyridin-3-yl)-2-oxoacetate
CCOC(=O)C(=O)C1=CN=C(C=C1)Cl
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
902837-55-2
C9H8ClNO3
213.617
MFCD07781188
HRFPVENOVYSKHE-UHFFFAOYSA-N
ethyl 6-chloro-3-pyridylglyoxylate, ethyl 2-6-chloropyridin-3-yl-2-oxoacetate, ethyl 2-chloro-5-pyridylglyoxylate, ethyl 6-chloropyridin-3-yl oxo acetate, ethyl6-chloro-3-pyridylglyoxylate, ethyl 6-chloro-3-pyridinyl oxoacetate, 6-chloro-3-pyridylglyoxylic acid ethyl ester, ethyl 2-6-chloro 3-pyridyl-2-oxoacetate
42553168
ethyl 2-(6-chloropyridin-3-yl)-2-oxoacetate
CCOC(=O)C(=O)C1=CN=C(C=C1)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 213.62
Percent Purity 97%
Chemical Name or Material Ethyl 2-chloro-5-pyridylglyoxylate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'