missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-amino-5-nitrothiophene-3-carboxylate, 95%, Thermo Scientific™

Catalog Number 15428178
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
42783-04-0
C7H8N2O4S
216.211
MFCD01566377
UXDKVBJDZDRIKL-UHFFFAOYSA-N
2-amino-3-ethoxycarbonyl-5-nitrothiophene, 3-thiophenecarboxylicacid, 2-amino-5-nitro-, ethyl ester, 2-amino-3-carbethoxy-5-nitrothiophene;, 2-amino-3-carboethoxy-5-nitrothiophene;, 2-amino-3-ethoxycarbonyl-5-nitrothiophene;, 2-amino-5-nitro-3-ethoxycarbonyl-thiophene, 2-amino-5-nitrothiophen-3-carbonsaeureethylester, 2-amino-5-nitro-3-thiophenecarboxylic acid ethyl ester, 2-amino-5-nitro-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid,2-amino-5-nitro-,ethyl ester
2737608
ethyl 2-amino-5-nitrothiophene-3-carboxylate
CCOC(=O)C1=C(SC(=C1)[N+](=O)[O-])N
This item is not returnable. View return policy
42783-04-0
C7H8N2O4S
216.211
MFCD01566377
UXDKVBJDZDRIKL-UHFFFAOYSA-N
2-amino-3-ethoxycarbonyl-5-nitrothiophene, 3-thiophenecarboxylicacid, 2-amino-5-nitro-, ethyl ester, 2-amino-3-carbethoxy-5-nitrothiophene;, 2-amino-3-carboethoxy-5-nitrothiophene;, 2-amino-3-ethoxycarbonyl-5-nitrothiophene;, 2-amino-5-nitro-3-ethoxycarbonyl-thiophene, 2-amino-5-nitrothiophen-3-carbonsaeureethylester, 2-amino-5-nitro-3-thiophenecarboxylic acid ethyl ester, 2-amino-5-nitro-thiophene-3-carboxylic acid ethyl ester, 3-thiophenecarboxylicacid,2-amino-5-nitro-,ethyl ester
2737608
ethyl 2-amino-5-nitrothiophene-3-carboxylate
CCOC(=O)C1=C(SC(=C1)[N+](=O)[O-])N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 1 g
Formula Weight 216.21
Percent Purity 95%
Physical Form Solid
Chemical Name or Material Ethyl 2-amino-5-nitrothiophene-3-carboxylate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'