missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EPPS, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
EPPS is used as a biological buffer with a useful pH range of 7.3-8.7 & pKa=8.0 (25°C). It can be used in agrochemical, pharmaceutical and dyestuff field etc.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
EPPS is used as a biological buffer with a useful pH range of 7.3-8.7 & pKa=8.0 (25°C). It can be used in agrochemical, pharmaceutical and dyestuff field etc.
Solubility
Soluble in water. 25.2 g/L (20°C)
Notes
Store at room temperature. Incompatible with oxidizing agents. Store away from extreme heat.
EPPS is used as a biological buffer with a useful pH range of 7.3-8.7 & pKa=8.0 (25°C). It can be used in agrochemical, pharmaceutical and dyestuff field etc.
Solubility
Soluble in water. 25.2 g/L (20°C)
Notes
Store at room temperature. Incompatible with oxidizing agents. Store away from extreme heat.
Specifications
Melting Point | 242°C (decomposition) |
Quantity | 25 g |
Beilstein | 3957385 |
Solubility Information | Soluble in water. 25.2g/L (20°C) |
Formula Weight | 252.33 |
Percent Purity | 99% |
Chemical Name or Material | EPPS |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'