Learn More
Sartorius Entris™ II Advanced Precision Balances with Country-Specific Verification
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
Brand:  Sartorius BCA623I-1CFR
Additional Details : Weight : 5.90000kg
 View more versions of this product
Product Code. 16655683
  • 3620.00 € / Each
Estimated Shipment: 29-12-2021
Add to basket
Description
Description
  • Entris™ II Advanced Balances - Quality, Value, and Consistency
  • For accurate weighing results that can be trusted time after time
  • Simplifies balance leveling with built-in, real-time level support
  • Increases reliability of weighing results using integrated protection systems
  • Controlled access to balance settings with user management
  • Gapless documentation thanks to CalAuditTrail
Specifications
Specifications
Verified (France Country-Specific)
Intuitive Graphic Touch Technology
D-sub connector, 9-pin; USB UTL; USB Device
1 mg
Glass, PBT, Stainless Steel
100/240 V (± 10%)
50/60 Hz
0.2 A
CE Certificate
Electronic Precision Balance
≤1.0 seconds
620 g
Videos
SDS
missing translation for 'documents'
Product Certifications
Special Offers
Special Offers