Learn More
Sartorius Entris™ II Advanced Analytical Balances with Country-Specific Verification
Over 38 different models offer various benefits such as real-time level support, integrated protection systems, CalAuditTrail, a graphic touch display, and 13 built-in application programs
Brand:  Sartorius BCA224I-1CFR
Additional Details : Weight : 5.90000kg
 View more versions of this product
Product Code. 16645653
  • 4120.00 € / Each
Estimated Shipment: 29-12-2021
Add to basket
Description
Description
  • Entris™ II Advanced balances - Quality, Value, and Consistency
  • For accurate weighing results that can be trusted time after time
  • Simplifies balance leveling with built-in real-time level support
  • Increases reliability of weighing results using integrated protection systems
  • Controlled access to balance settings with user management
  • Gapless documentation thanks to CalAuditTrail
Specifications
Specifications
Verified (France Country Specific)
220 g
0.2 A
0.1 mg
≤1.5 sec.
D-sub connector, 9-pin; USB UTL; USB Device
50/60 Hz
Intuitive Graphic Touch Technology
CE Certificate
Glass, PBT, Stainless Steel
100/240 V (±10%)
Electronic Analytical Balance