Learn More
Merck Millipore RoClean™ Cleaning Pouches for Water Purification Systems
Keep your purification system clean with easy-to-use RoClean pouches.
POA
Specifications
For Use With (Equipment) Elix™, Milli-RO™, Milli-RX™ and Millipore™ RiOs™ water purification system
View More Specs
Description
Description
  • For cleaning RO membrane in Elix™, Milli-RO™, Milli-RX™, and low-volume Millipore™ RiOs™ Water Purification Systems
  • Available in acidic (ROClean A) and basic (ROClean B) formulas
†For details, see Catalog 2004/05.
Specifications
Specifications
Elix™, Milli-RO™, Milli-RX™ and Millipore™ RiOs™ water purification system