Learn More
Macherey-Nagel™ EC Analytical HPLC Column
Shop All Macherey-Nagel Products
Click to view available options
Stationary Phase:
BSA-7
Permethylated α-cyclodextrin
Permethylated β-cyclodextrin
Permethylated γ-cyclodextrin
β-cyclodextrin
Unit Size:
Each
Specifications
Specifications
| Column Chemistry | Silica Base |
| Column Format | Analytical Column |
| Diameter (Metric) | 4 mm |
| For Use With | For HPLC Enantiomer Separation |
| Length (Metric) | 200 mm |
| Mobile Phase Compatibility | CH3OH - 0.1 % TEAA pH 4 (65:35) |
| Particle Size | 5 μm |
| Pore Size | 100 Å |
| USP Type | L45 |
| Stationary Phase | Permethylated β-cyclodextrin |
| Show More |