missing translation for 'onlineSavingsMsg'
Learn More

E6130, MedChemExpress

Product Code. p-200100829
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476837

Brand: Medchem Express HY107456

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress E6130 is an orally active and highly selective CX3CR1 modulator, that may be effective for treatment of inflammatory bowel disease.

E6130 is an orally active and highly selective CX3CR1 modulator, that may be effective for treatment of inflammatory bowel disease.

Specifications

Chemical Name or Material E6130
CAS 1427058-33-0
Color White
Purity Grade Notes Research
Molecular Formula C28H37ClF3N3O3
Quantity 5 mg
Solubility Information DMSO : ≥ 250 mg/mL (449.59 mM)
SMILES C[C@@H](C1)[C@@](C(NC2CCN(CC3=CCCCC3)CC2)=O)(CC(O)=O)CN1CC4=C(C(F)(F)F)C=CC=C4Cl
Molecular Weight (g/mol) 556.06
Formula Weight 556.06
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.