Learn More
Dysferlin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554541
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: CHYYYLPWGN VKPVVVLSSY WEDISHRIET QNQLLGIADR LEAGLEQVHL ALKAQCSTED VDSLVAQLTD ELIAGCSQPL GDIHETPSAT HLDQYLYQLR THHLSQITEA ALALKLGHSE LPAALEQAED WLLRLRALA Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 88%.
Dysferlin Polyclonal antibody specifically detects Dysferlin in Human samples. It is validated for Immunohistochemistry.Specifications
Dysferlin | |
Polyclonal | |
Unconjugated | |
DYSF | |
2310004N10Rik; AI604795; D6Pas3; Dysf; Dysferlin; dysferlin variant a; dysferlin_a; dystrophy-associated fer-1-like 1; dystrophy-associated fer-1-like protein; FER1L1; fer-1-like family member 1; fer-1-like protein 1; FLJ00175; FLJ90168; LGMD2B; limb girdle muscular dystrophy 2B (autosomal recessive); MMD1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8291 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75923 | |
DYSF | |
Recombinant protein corresponding to Human Dysferlin | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only