Learn More
3M™ 5000 Series Particulate Filters
Provides excellent particulate protection in the laboratory. 3M™ 5000 Series Particulate Filters are available in a range of protection levels.
Brand:  3M™ 7000037675
Additional Details : Weight : 0.18000kg
Product Code. 11773343
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Combines with the 6000 Series Gas Vapour cartridges to provide particulate and gas/vapour protection
  • Note: 3M 501 filter retainers needed to combine the 5000 and 6000 series
Specifications
Specifications
P3R
Dust Filter
20/Pk.
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products