Learn More
DWK Life Sciences DURAN™ Plastic Stopper, White Polyethylene, Octagonal flat head
Plastic stopper for flasks and bottles with glass ground joints
Brand: DWK Life Sciences 292040809
16.44 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Additional Details : Weight : 0.07200kg
Description
Stoppers with octagonal top made of polyethylene plastic. Conforms to DIN 12254. White with blue insert.
- Made from polyethylene
- Octagonal shaped head
Specifications
Storage of reagents, and chemicals | |
10/Pk. | |
No |
ST 24/29 | |
Polyethylene |