Learn More
DPP-IV-IN-1, MedChemExpress
Product Code. 16492558
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16492558
Brand: Medchem Express HYU00346
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress DPP-IV-IN-1 is a potent inhibitor of dipeptidyl peptidase IV (DPP-IV), a highly specific serine protease, with an IC50 of 4.6 nM.
DPP-IV-IN-1 is a potent inhibitor of dipeptidyl peptidase IV (DPP-IV), a highly specific serine protease, with an IC50 of 4.6 nM.
Specifications
Chemical Name or Material DPP-IV-IN-1
CAS 625110-37-4
Color White
Purity Grade Notes Research
Molecular Formula C11H18FN3O2
Quantity 5 mg
Solubility Information DMSO : 125 mg/mL (513.81 mM; Need ultrasonic)
SMILES N#C[C@H]1N(C(CNC(C)(C)CO)=O)C[C@@H](F)C1
Molecular Weight (g/mol) 243.28
Formula Weight 243.28
Percent Purity 98.79%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-sugar/lipid metabolism