missing translation for 'onlineSavingsMsg'
Learn More

Dowex™ 1X2 100-200 (Cl), Thermo Scientific Chemicals

Catalog Number p-7046415
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
9085-42-1
C29H34ClN
432.05
MFCD00132715
BBQMUEOYPPPODD-UHFFFAOYSA-M
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
16212807
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
[Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1
This item is not returnable. View return policy
9085-42-1
C29H34ClN
432.05
MFCD00132715
BBQMUEOYPPPODD-UHFFFAOYSA-M
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
16212807
1,4-bis(ethenyl)benzene;(4-ethenylphenyl)-trimethylazanium;styrene;chloride
[Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Odor Amine-like
Quantity 50 g
Chemical Name or Material Dowex™ 1X2 100-200 (Cl)

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'