Learn More
BRAND™ Double-Ended PTFE Magnetic Stirring Bars
Excellent centering
POA - 226.00 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
12025292
|
BRAND™
137930 |
37 mm |
226.00 €
Pack of 10 |
Estimated Shipment: 03-05-2024
Log in to see stock. |
|||||
10668881
|
BRAND™
137935 |
54 mm |
POA
Pack of 10 |
Estimated Shipment: 14-05-2024
Log in to see stock. |
|||||
Description
Specifications
8 mm | |
10 | |
Dumbbell |