missing translation for 'onlineSavingsMsg'
Learn More

Motic™ Gliding Stage, Circular, 2106K

Suitable for FBGG/2111/2112 base stands and used for smooth multidirectional movement. Motic™ Gliding Stage, Circular, 2106K is mounted onto the base plate of a plain or transmitted light stand.

Brand:  Motic™ 1101000600542

Additional Details : Weight : 0.01000kg

Product Code. 15495202

  • 95.00 € / Each
Estimated Shipment: 03-07-2024
to see stock.
Add to basket

Description

Description

  • Used for smooth multidirectional movement.
  • 360° rotatable gliding stage.
  • Stage is mounted onto the base plate of a plain or transmitted light stand.
Specifications

Specifications

Circular Gliding Stage
FBGG/2111/2112 base stands
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions