missing translation for 'onlineSavingsMsg'
Learn More

DL-Prolinol, 98%, Thermo Scientific Chemicals

Catalog Number p-7058155
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
498-63-5
C5H11NO
101.149
MFCD00601073
HVVNJUAVDAZWCB-UHFFFAOYSA-N
2-hydroxymethyl pyrrolidine, 2-pyrrolidinemethanol, prolinol, pyrrolidin-2-yl-methanol, dl-prolinol, 2-hydroxymethylpyrrolidine, pyrrolidin-2-yl methanol, 2-pyrrolidinylmethanol, pyrrolidine-2-methanol, 2-pyrrolidinylmethanol #
90908
CHEBI:59652
pyrrolidin-2-ylmethanol
C1CC(NC1)CO
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
498-63-5
C5H11NO
101.149
MFCD00601073
HVVNJUAVDAZWCB-UHFFFAOYSA-N
2-hydroxymethyl pyrrolidine, 2-pyrrolidinemethanol, prolinol, pyrrolidin-2-yl-methanol, dl-prolinol, 2-hydroxymethylpyrrolidine, pyrrolidin-2-yl methanol, 2-pyrrolidinylmethanol, pyrrolidine-2-methanol, 2-pyrrolidinylmethanol #
90908
CHEBI:59652
pyrrolidin-2-ylmethanol
C1CC(NC1)CO

DL-Prolinol is used as asymmetric catalysts in proline organ catalysis reactions, and it is also used in many pharmaceutical, biotechnological applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
DL-Prolinol is used as asymmetric catalysts in proline organ catalysis reactions, and it is also used in many pharmaceutical, biotechnological applications.

Solubility
Soluble in ethanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Refractive Index 1.484
Boiling Point 76°C (2mmHg)
Quantity 5 g
Solubility Information Soluble in ethanol.
Formula Weight 101.15
Percent Purity 98%
Chemical Name or Material DL-Prolinol

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'