missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DL-Ethionine, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
DL-Ethionine is used as a dietary supplement to accelerate cholangiocarcinogenesis in vivo. It is also used to induce oxidative stress in liver to study the levels and activities of anti-oxidative enzymes and compounds such as glutathione.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | ∼265°C (decomposition) |
Odor | Odorless |
Quantity | 1 g |
Beilstein | 1722529 |
Merck Index | 14,3738 |
Solubility Information | Soluble in water. |
IUPAC Name | 2-amino-4-(ethylsulfanyl)butanoic acid |
Formula Weight | 163.24 |
Percent Purity | 98% |
Chemical Name or Material | DL-Ethionine |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'