missing translation for 'onlineSavingsMsg'
Learn More

Dichloromethane, Puriss., meets analytical spec. of Ph. Eur., NF, ≥99% (GC), Honeywell Riedel-de Haën™
BIOPHARMA

Catalog Number p-7120375
88.59 EUR valid until 2024-03-31
Use promo code "21663" to get your promotional price.


Click to view available options
Quantity:
5 L
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Aluminum Bottle
Glass Bottle
75-09-2
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
6344
CHEBI:15767
dichloromethane
ClCCl
75-09-2
CH2Cl2
84.93
MFCD00000881
YMWUJEATGCHHMB-UHFFFAOYSA-N
methylene chloride, methylene dichloride, methane, dichloro, methylene bichloride, methane dichloride, solaesthin, solmethine, freon 30, narkotil, aerothene mm
6344
CHEBI:15767
dichloromethane
ClCCl

Puriss., meets analytical specification of Ph. Eur., NF,≥99% (GC)

  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY
Melting Point -97°C
Color Colorless
UN Number UN1593
Beilstein 1730800
Formula Weight 84.93g/mol
Percent Purity ≥99% (GC)
Grade puriss.
Density 1.318 to 1.322g/mL at 25°C, 1.325g/mL at 25°C
Boiling Point 39°C to 41°C
Quantity 5 L
Explosion Limit 0.22
Ignition Point 1223°F
Packaging Aluminum Bottle
Physical Form Liquid
Vapor Pressure 24.45psi (55°C), 6.83psi (20°C)
Chemical Name or Material Dichloromethane, Meets analytical specification of Ph.Eur.NF
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Dichloromethane
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'