Learn More
Diaion™ PK208(Na), ion exchange resin, 4% cross-linked, strongly acidic porous type, 1.2 meq/ml on PS-DVB, Thermo Scientific™
Brand: Thermo Scientific Alfa Aesar 046476.A1
Additional Details : Weight : 1.00000kg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Used in demineralization, decolorization, metal recovery, pharmaceuticals, amino acid separation
Specifications
300-1180μm Beads | |
4% cross-linked, strongly acidic porous type, 1.2meq/mL on PS-DVB | |
300 to 1180μm |
Diaion™ PK208 (Na), ion exchange resin | |
Odorless | |
1 kg |