missing translation for 'onlineSavingsMsg'
Learn More

Di-n-hexyl phthalate, 97%, Thermo Scientific Chemicals

Product Code. p-7020585
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 11444314

Brand: Thermo Scientific Alfa Aesar A12593.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Phthalates are by far the most widely used plasticizers, primarily to make soft and flexible polyvinyl chloride (PVC) for the applications in the industry of automotive, building & construction material, cable, flooring, medical device and toys. Minor quantity of phthalates are used in adhesives, caulk, sealants, paint to improve work performance. Small molecule phthalates are used as solvents in perfumes to provide longer linger longer and in nail polish to prevent chipping.

Solubility
Soluble in water (0.24 mg/L).

Notes
Store at 4°C. Avoid excessive heat and light. Reactive towards oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 84-75-3
Molecular Formula C20H30O4
Molecular Weight (g/mol) 334.46
MDL Number MFCD00043720
InChI Key KCXZNSGUUQJJTR-UHFFFAOYSA-N
Synonym dihexyl phthalate, di-n-hexyl phthalate, di-n-hexylphthalate, phthalic acid, dihexyl ester, 1,2-benzenedicarboxylic acid, dihexyl ester, phthalic acid di-n-hexyl ester, phthalic acid dihexyl ester, n-dihexyl phthalate, bis n-hexyl phthalate, dihexyl 1,2-benzenedicarboxylate
PubChem CID 6786
ChEBI CHEBI:34678
IUPAC Name dihexyl benzene-1,2-dicarboxylate
SMILES CCCCCCOC(=O)C1=CC=CC=C1C(=O)OCCCCCC

Specifications

Density 1.01
Boiling Point 185°C to 187°C (0.5 mmHg)
Refractive Index 1.487
Quantity 10 g
UN Number UN3082
Solubility Information Soluble in water (0.24mg/L).
Formula Weight 334.46
Percent Purity 97%
Chemical Name or Material Di-n-hexyl phthalate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.