missing translation for 'onlineSavingsMsg'
Learn More

Di-n-butyl maleate, 96%, Thermo Scientific™

Product Code. p-7039736
Click to view available options
Quantity:
0.25 L
1 L
Unit Size:
0.25 L
1 L
This item is not returnable. View return policy

Product Code. 15465338

Brand: Thermo Scientific Alfa Aesar H59123.K1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-76-0
Molecular Formula C12H20O4
Molecular Weight (g/mol) 228.29
MDL Number MFCD00009447,MFCD00065141
InChI Key JBSLOWBPDRZSMB-FPLPWBNLSA-N
Synonym dibutyl maleate, butyl maleate, staflex dbm, rc comonomer dbm, maleic acid, dibutyl ester, di-n-butyl maleate, 2-butenedioic acid z-, dibutyl ester, 2-butenedioic acid, dibutyl ester, maleic acid dibutyl ester, 2-butenedioic acid 2z-, dibutyl ester
PubChem CID 5271569
IUPAC Name dibutyl (Z)-but-2-enedioate
SMILES CCCCOC(=O)\C=C/C(=O)OCCCC

Specifications

Density 0.988
Boiling Point 281°C
Flash Point 138°C (280°F)
Linear Formula CH3(CH2)3OCOCH=CHCOO(CH2)3CH3
Refractive Index 1.445
Quantity 0.25 L
Beilstein 1726634
Formula Weight 228.28
Percent Purity 96%
Chemical Name or Material Di-n-butyl maleate
compliance-icons compliance-icons compliance-icons
Signal Word
  • Warning
Hazard Category
  • ACUTE AQUATIC Acute 1
  • Skin sensitiser Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H373-May cause damage to organs.
  • H400-Very toxic to aquatic life.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P314-Get medical advice/attention if you feel unwell.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.