missing translation for 'onlineSavingsMsg'
Learn More

Dextran sulfate sodium salt, MW ca >500,000, Thermo Scientific Chemicals

Catalog Number p-7042508
Click to view available options
Quantity:
5 g
50 g
100 g
1 kg
9011-18-1
(C6H7O5)m(C6H7O5)n
40,000
MFCD00081551
BLFLLBZGZJTVJG-UHFFFAOYSA-N
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
2337
CHEBI:116735
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]
9011-18-1
(C6H7O5)m(C6H7O5)n
40,000
MFCD00081551
BLFLLBZGZJTVJG-UHFFFAOYSA-N
benzocaine, ethyl aminobenzoate, ethyl p-aminobenzoate, americaine, anesthesin, anaesthesin, ethoform, norcaine, orthesin, parathesin
2337
CHEBI:116735
*-OC[C@H]1O[C@H](OC[C@H]2O[C@H](-*)[C@H](O[*])[C@@H](O[*])[C@@H]2O[*])[C@H](O[*])[C@@H](O-*)[C@@H]1O[*]

Dextran sulfate sodium is used to study colitis (inflammatory bowel disease) as well as an antiviral agent. It functions as a nonsurfactant, suspending agent with gel forming and viscosity controlling properties. It can be used in gels and moisturizers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White to Yellow
Physical Form Powder
Merck Index 14,2951
Solubility Information Soluble in water.
Quantity 5 g
IUPAC Name Dextran sulfate sodium salt
Formula Weight >≈500 kd
Odor Phenol-like
Chemical Name or Material Dextran sulfate sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'