Learn More
DEPC-treated WaterAbnova™
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Description
- The DEPC-treated Water was prepared by 0.1 % DEPC-treatment and autoclave of highly purified water.
- The product was confirmed to be RNase-free and DNase free.
- Apperance is clear liquid.
Specifications
Quantity | 5 mL |