missing translation for 'onlineSavingsMsg'
Learn More

Decyl decanoate, 98%, Thermo Scientific Chemicals

Catalog Number p-7077659
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1654-86-0
C20H40O2
312.54
XAKXZZPEUKNHMA-UHFFFAOYSA-N
decanoic acid, decyl ester, decanoic acid decyl ester, unii-b88b7acl5l, b88b7acl5l, n-capric acid n-decyl ester, capryl caprate, acmc-209dsk, decanoic acid, decylester
74247
decyl decanoate
CCCCCCCCCCOC(=O)CCCCCCCCC
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
1654-86-0
C20H40O2
312.54
XAKXZZPEUKNHMA-UHFFFAOYSA-N
decanoic acid, decyl ester, decanoic acid decyl ester, unii-b88b7acl5l, b88b7acl5l, n-capric acid n-decyl ester, capryl caprate, acmc-209dsk, decanoic acid, decylester
74247
decyl decanoate
CCCCCCCCCCOC(=O)CCCCCCCCC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Density 0.8590g/mL
Boiling Point 174.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula CH3(CH2)8CO2(CH2)9CH3
Quantity 25 mL
Specific Gravity 0.859
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 312.54
Percent Purity 98%
Chemical Name or Material Decyl decanoate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'