missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decyl beta-D-glucopyranoside, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Non-ionic detergent used for solubilizing membrane proteins and in the development of sugar-based surfactant solutions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Non-ionic detergent used for solubilizing membrane proteins and in the development of sugar-based surfactant solutions.
Specifications
Color | White |
Quantity | 1 g |
Beilstein | 85089 |
Solubility Information | Soluble in methanol |
Formula Weight | 320.42 |
Physical Form | Powder |
Chemical Name or Material | Decyl beta-D-glucopyranoside |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'