Learn More
Macherey-Nagel™ Decapping Head
445.00 €
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Price | Quantity & Availability | |||||
12808605
|
Macherey-Nagel™
735811 |
For Electronic High Power Crimping Tool 735700 and 11mm Crimp Caps |
445.00 €
Each |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12818605
|
Macherey-Nagel™
735820 |
For Electronic High Power Crimping Tool 735700 and 20mm Crimp Caps |
445.00 €
Each |
Estimated Shipment: 06-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Decapping Head | |
1 Each |