Learn More
DCAF8 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA555548
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EDKDSDSMED TGHYSINDEN RVHDRSEEEE EEEEEEEEEQ PRRRVQRKRA NRDQDSSDDE RALEDWVS Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 82%.
DCAF8 Polyclonal antibody specifically detects DCAF8 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
DCAF8 | |
Polyclonal | |
Unconjugated | |
DCAF8 | |
AA408877; AA433120; AA792214; AU018973; C76055; D1Dau35e; D1Ucla4; Dcaf8; DDB1 and CUL4 associated factor 8; DDB1- and CUL4-associated factor 8; GAN2; H326; WD repeat domain 42A; WD repeat-containing protein 42A; Wdr42a | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
50717 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q5TAQ9 | |
DCAF8 | |
Recombinant protein corresponding to Human DCAF8 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only