missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Threonine, 98%

Catalog Number p-3776665
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass Bottle
This item is not returnable. View return policy
632-20-2
C4H9NO3
119.12
MFCD00064269
AYFVYJQAPQTCCC-STHAYSLISA-N
d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dth
69435
CHEBI:16398
(2R,3S)-2-amino-3-hydroxybutanoic acid
C[C@H](O)[C@@H](N)C(O)=O
This item is not returnable. View return policy
632-20-2
C4H9NO3
119.12
MFCD00064269
AYFVYJQAPQTCCC-STHAYSLISA-N
d-threonine, h-d-thr-oh, 2r,3s-2-amino-3-hydroxybutanoic acid, threonine, d, r-threonine, d-threonin, d-2-amino-3-hydroxybutyric acid, 2r,3s-2-amino-3-hydroxybutyric acid, d-thr, dth
69435
CHEBI:16398
(2R,3S)-2-amino-3-hydroxybutanoic acid
C[C@H](O)[C@@H](N)C(O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Ammonium 200ppm max.
Arsenic (As) 1ppm max.
Color White
Chlorine (Cl) 200ppm max.
Heavy Metals (as Pb) 10ppm max.
Assay Percent Range 98%
Infrared Spectrum Authentic
Linear Formula CH3CH(OH)CH(NH)COH
Iron (Fe) 10ppm max.
Loss on Drying 0.3% max.
Other Amino Acids (TLC) Passes Test
Packaging Glass Bottle
Specific Rotation +27° to +29° (20°C, 589nm) (c=6, water)
Sulfate 300ppm max.
Formula Weight 119.12
Transmittance 98% min. (c=5, H2O, 430nm) 1cm cell
Percent Purity ≥97.5%
Sulfated Ash 0.1% max.
Specific Rotation Condition +28° (20°C c=6,water)
Quantity 5 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D-Threonine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'