missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Phenylalanine tert-butyl ester hydrochloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
D-Phenylalanine tert-butyl ester hydrochloride is used as a fine chemical intermediate, agrochemical &pharmceutical intermediate. It has anti - static and anti- corrosion properties.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Phenylalanine tert-butyl ester hydrochloride is used as a fine chemical intermediate, agrochemical &pharmceutical intermediate. It has anti - static and anti- corrosion properties.
Solubility
Soluble in water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
D-Phenylalanine tert-butyl ester hydrochloride is used as a fine chemical intermediate, agrochemical &pharmceutical intermediate. It has anti - static and anti- corrosion properties.
Solubility
Soluble in water.
Notes
Heat sensitive. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications
Solubility Information | Soluble in water. |
Formula Weight | 257.76 |
Percent Purity | 98% |
Quantity | 250 mg |
Physical Form | Solid |
Chemical Name or Material | D-Phenylalanine tert-butyl ester hydrochloride |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'