missing translation for 'onlineSavingsMsg'
Learn More

D-myo-Inositol-2,5,6-trisphosphate sodium salt, 98%, Thermo Scientific™

Catalog Number 15418609
Click to view available options
Quantity:
0.1 mg
0.5 mg
This item is not returnable. View return policy
MFCD26967929
Ins(2,5,6)-P3 sodium salt; 2,5,6-IP3 sodium salt
This item is not returnable. View return policy
MFCD26967929
Ins(2,5,6)-P3 sodium salt; 2,5,6-IP3 sodium salt

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Color White
Quantity 0.5 mg
Formula Weight 486.04
Percent Purity 98%
Physical Form Lyophilized Powder
Chemical Name or Material D-myo-Inositol-2,5,6-trisphosphate sodium salt
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'