missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Isoascorbic acid sodium salt monohydrate, 99%, Thermo Scientific™

Catalog Number p-7032272
Click to view available options
Quantity:
100 g
2500 g
This item is not returnable. View return policy
63524-04-9
C6H9NaO7
216.121
MFCD03788863
NKESWEQTVZVSSF-DMWQRSMXSA-M
sodium erythorbate monohydrate, sodium d-isoascorbate monohydrate, d-isoascorbic acid sodium salt monohydrate, d-isoascorbic acid sodium salt monohydrate; sodium erythorbate monohydrate, 5r-5-1,2-dihydroxyethyl-3,4-dihydroxy-5-hydrofuran-2-one, sodium salt, hyd rate, 5r-5-1r-1,2-dihydroxyethyl-4-hydroxy-3-sodiooxy-5h-furan-2-one hydrate, sodium 5r-5-1r-1,2-dihydroxyethyl-4-hydroxy-2-oxo-5h-furan-3-olate hydrate, sodium 5r-5-1,2-dihydroxyethyl-4-hydroxy-2-oxo-2,5-dihydrofuran-3-olate-water 1/1/1 non-preferred name
54691414
sodium;(2R)-2-[(1R)-1,2-dihydroxyethyl]-3-hydroxy-5-oxo-2H-furan-4-olate;hydrate
C(C(C1C(=C(C(=O)O1)[O-])O)O)O.O.[Na+]
This item is not returnable. View return policy
63524-04-9
C6H9NaO7
216.121
MFCD03788863
NKESWEQTVZVSSF-DMWQRSMXSA-M
sodium erythorbate monohydrate, sodium d-isoascorbate monohydrate, d-isoascorbic acid sodium salt monohydrate, d-isoascorbic acid sodium salt monohydrate; sodium erythorbate monohydrate, 5r-5-1,2-dihydroxyethyl-3,4-dihydroxy-5-hydrofuran-2-one, sodium salt, hyd rate, 5r-5-1r-1,2-dihydroxyethyl-4-hydroxy-3-sodiooxy-5h-furan-2-one hydrate, sodium 5r-5-1r-1,2-dihydroxyethyl-4-hydroxy-2-oxo-5h-furan-3-olate hydrate, sodium 5r-5-1,2-dihydroxyethyl-4-hydroxy-2-oxo-2,5-dihydrofuran-3-olate-water 1/1/1 non-preferred name
54691414
sodium;(2R)-2-[(1R)-1,2-dihydroxyethyl]-3-hydroxy-5-oxo-2H-furan-4-olate;hydrate
C(C(C1C(=C(C(=O)O1)[O-])O)O)O.O.[Na+]

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 100 g
Optical Rotation +97° (c=10 in Water)
Formula Weight 216.12 (198.11 Anhydrous)
Percent Purity 99%
Chemical Name or Material D-(+)-Isoascorbic acid sodium salt monohydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'