missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ D-(+)-Isoascorbic acid sodium salt monohydrate, 99%

Brand:  Alfa Aesar™ H33670.0E

Additional Details : CAS : 63524-04-9 Weight : 2.50000kg

Quantity 2500g

 View more versions of this product

Product Code. 15418575

  • 433.00 € / 2500g


Estimated Shipment: 29-06-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

63524-04-9
216.121
NKESWEQTVZVSSF-DMWQRSMXSA-M
54691414
C(C(C1C(=C(C(=O)O1)[O-])O)O)O.O.[Na+]
C6H9NaO7
MFCD03788863
sodium erythorbate monohydrate, sodium d-isoascorbate monohydrate, d-isoascorbic acid sodium salt monohydrate, d-isoascorbic acid sodium salt monohydrate; sodium erythorbate monohydrate, 5r-5-1,2-dihydroxyethyl-3,4-dihydroxy-5-hydrofuran-2-one, sodium salt, hyd rate, 5r-5-1r-1,2-dihydroxyethyl-4-hydroxy-3-sodiooxy-5h-furan-2-one hydrate, sodium 5r-5-1r-1,2-dihydroxyethyl-4-hydroxy-2-oxo-5h-furan-3-olate hydrate, sodium 5r-5-1,2-dihydroxyethyl-4-hydroxy-2-oxo-2,5-dihydrofuran-3-olate-water 1/1/1 non-preferred name
sodium;(2R)-2-[(1R)-1,2-dihydroxyethyl]-3-hydroxy-5-oxo-2H-furan-4-olate;hydrate

Specifications

D-(+)-Isoascorbic acid sodium salt monohydrate
2500g
C6H9NaO7
sodium erythorbate monohydrate, sodium d-isoascorbate monohydrate, d-isoascorbic acid sodium salt monohydrate, d-isoascorbic acid sodium salt monohydrate; sodium erythorbate monohydrate, 5r-5-1,2-dihydroxyethyl-3,4-dihydroxy-5-hydrofuran-2-one, sodium salt, hyd rate, 5r-5-1r-1,2-dihydroxyethyl-4-hydroxy-3-sodiooxy-5h-furan-2-one hydrate, sodium 5r-5-1r-1,2-dihydroxyethyl-4-hydroxy-2-oxo-5h-furan-3-olate hydrate, sodium 5r-5-1,2-dihydroxyethyl-4-hydroxy-2-oxo-2,5-dihydrofuran-3-olate-water 1/1/1 non-preferred name
C(C(C1C(=C(C(=O)O1)[O-])O)O)O.O.[Na+]
216.121
216.12 (198.11 Anhydrous)
+97° (c=10 in Water)
63524-04-9
MFCD03788863
NKESWEQTVZVSSF-DMWQRSMXSA-M
sodium;(2R)-2-[(1R)-1,2-dihydroxyethyl]-3-hydroxy-5-oxo-2H-furan-4-olate;hydrate
54691414
99%
Videos
SDS
Documents
Certificates