missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadecane, 98%

Product Code. p-7024554
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11424017

Brand: Thermo Scientific Alfa Aesar A19416.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 295-48-7 | C15H30 | 210.405 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 295-48-7
Molecular Formula C15H30
Molecular Weight (g/mol) 210.405
MDL Number MFCD00039424
InChI Key SRONXYPFSAKOGH-UHFFFAOYSA-N
Synonym acmc-209h9p
PubChem CID 67525
IUPAC Name cyclopentadecane
SMILES C1CCCCCCCCCCCCCC1

Specifications

Density 0.805
Quantity 1 g
Formula Weight 210.4
Percent Purity 98%
Chemical Name or Material Cyclopentadecane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.