Learn More
Custom PreAmp Pools
Custom PreAmp Pools
Brand:  ABI Applied Biosystems 4459657
Additional Details : Weight : 0.00100kg
 View more versions of this product
Product Code. 15524344
  • 442.00 € / 1mL
Estimated Shipment: 19-04-2024
to see stock.
Add to basket
For Research Use Only. All usage must comply with product instructions.
Description
Description
Custom miRNA array designs can hold from 12 to 380 assays, and 1 to 8 samples on each array design. Ten formats are offered, available only with microfluidics card design. Use Custom PreAmp pools for samples with limited miRNA amounts.
Specifications
Specifications
Custom TaqMan™ miRNA Pre-Amp Pool
Room Temperature
1 mL
Format 24
1 to 8