Learn More
Simport™ Scientific Cryovial™ External Thread with Lip Seal Cryogenic Vials
Designed for storing biological material, human or animal cells, at temperatures as low as -196°C. Simport™ Scientific Cryovial™ External Thread with Lip Seal Cryogenic Vials should be used only in the gas phase of liquid nitrogen.
POA
Specifications
Autoclavable Autoclavable
Material Polypropylene
Thread Style External
Sterility Sterile
Color Natural
View More Specs
Description
Description
  • Made of specially formulated polypropylene
  • Cap features a long skirt and for one hand aseptic methods
  • Fast cap thread design for tightening or removal with a mere 1 1/4 turn and inside thread design will not contribute to contamination
  • Lip inside cap ensures leakproof seal even at low temperatures
  • Provided with white marking area for sample identification
  • Color coded by the use of a CAPINSERT™
  • Sterilized by gamma radiation
  • Packaged in unique tamperproof, resealable, safety-lock bags
Certifications
CE Marked; RNase, DNase, Pyrogen and DNA-free
Specifications
Specifications
Autoclavable
External
Natural
Polypropylene
Sterile
Videos
SDS
Documents
Documents
Product Certifications
For in vitro diagnostic (IVD) use only.