Learn More
Cole-Parmer™ CPC™ All-Plastic Quick Disconnect Barbed Inserts
These couplings add ease of use and security to your most critical fluid handling applications
Brand:  Cole-Parmer™ 31311-73
Additional Details : Weight : 0.01000kg
Product Code. 15427163
  • 143.00 € / Pack of 5
Estimated Shipment: 19-06-2024
to see stock.
Add to basket
Description
Description
  • All-plastic quick-disconnect couplings require a standard fitting body and standard insert or large fitting body and large insert (order separately).
  • Simply press the thumb latch to disconnect fitting halves.
  • Available in polycarbonate (PC), Acetal and polysulfone (PSF).
Specifications
Specifications
All-Plastic Quick Disconnect Barbed Inserts; PS, 1/2 in. Tube ID
Quick-Disconnect x Barb
Polysulfone
5/Pk.
Autoclave, Ethylene Oxide (ETO) or Gamma