Learn More
Hach™ Copper Reagent Powder Pillow Set
Set of powder reagents in individual packets for Copper determination. Hach™ Copper Reagent Powder Pillow Set includes Copper Masking Reagent, Porphyrin 1 Reagent and Porphyrin 2 Reagent. Replacement porphyrin reagents available separately for replenishment after preparing blanks.
Brand: Hach™ 2603300
Additional Details : CAS : 50-81-7 Weight : 0.25000kg
Description
Set of powder reagents in individual packets for Copper determination. Hach™ Copper Reagent Powder Pillow Set includes Copper Masking Reagent, Porphyrin 1 Reagent and Porphyrin 2 Reagent. Replacement porphyrin reagents available separately for replenishment after preparing blanks.
- Ready-to-use, premeasured powder reagents in individual sealed packets
- For Copper determination by the Porphyrin Method (Hach Method 8143)
- Contains sets of copper masking and porphyrin 1/2 reagent powder pillows for 100 tests
- Porphyrin 1 and Porphyrin 2 powder pillows available separately
Specifications
Copper | |
Copper determination | |
Copper Masking Reagent Powder Pillows, Porphyrin 1 Reagent Powder Pillows and Porphyrin 2 Reagent Powder Pillows | |
1 Set | |
10 mL | |
Reagent set |
Powdered Reagent | |
DR 6000, DR/4000, DR 3900, DR 2800, ,DR 2700, DR/2010, DR/890 | |
100 | |
2 to 210μg/L Cu | |
Porphyrin |