Learn More
Hach™ Copper Reagent Powder Pillow Set
Set of powder reagents in individual packets for Copper determination. Hach™ Copper Reagent Powder Pillow Set includes Copper Masking Reagent, Porphyrin 1 Reagent and Porphyrin 2 Reagent. Replacement porphyrin reagents available separately for replenishment after preparing blanks.
Brand:  Hach™ 2603300
Additional Details : CAS : 50-81-7 Weight : 0.25000kg
 View more versions of this product
Product Code. 10432604
  • 525.00 € / Each
Estimated Shipment: 13-05-2024
to see stock.
Add to basket
Description
Description
Set of powder reagents in individual packets for Copper determination. Hach™ Copper Reagent Powder Pillow Set includes Copper Masking Reagent, Porphyrin 1 Reagent and Porphyrin 2 Reagent. Replacement porphyrin reagents available separately for replenishment after preparing blanks.
  • Ready-to-use, premeasured powder reagents in individual sealed packets
  • For Copper determination by the Porphyrin Method (Hach Method 8143)
  • Contains sets of copper masking and porphyrin 1/2 reagent powder pillows for 100 tests
  • Porphyrin 1 and Porphyrin 2 powder pillows available separately
Specifications
Specifications
Copper
Copper determination
Copper Masking Reagent Powder Pillows, Porphyrin 1 Reagent Powder Pillows and Porphyrin 2 Reagent Powder Pillows
1 Set
10 mL
Reagent set
Powdered Reagent
DR 6000, DR/4000, DR 3900, DR 2800, ,DR 2700, DR/2010, DR/890
100
2 to 210μg/L Cu
Porphyrin