missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) Sulfate, Pentahydrate, ACS, 99.0 to 100.5%, MilliporeSigma™

Encompass_Preferred
Click to view available options
Quantity:
250g
This item is not returnable. View return policy
7758-99-8
CuH10O9S
249.68
MFCD00149681
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
24463
CHEBI:31440
copper(2+) pentahydrate sulfate
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
7758-99-8
CuH10O9S
249.68
MFCD00149681
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
24463
CHEBI:31440
copper(2+) pentahydrate sulfate
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
Color Blue
Quantity 250g
Assay Percent Range 99.0 to 100.5%
Linear Formula CuSO4 · 5H2O
Formula Weight 249.68
Physical Form Crystals
Grade ACS/EP/ISO/Reagent
Density 2.284g/cm3 (20°C)
Chemical Name or Material Copper(II) Sulfate, Pentahydrate, EMSURE™, For Analysis