missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) sulfate pentahydrate, 98%, extra pure, Thermo Scientific Chemicals

Catalog Number p-3734770
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7758-99-8
CuH10O9S
249.68
MFCD00149681
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
24463
CHEBI:31440
copper;sulfate;pentahydrate
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
7758-99-8
CuH10O9S
249.68
MFCD00149681
JZCCFEFSEZPSOG-UHFFFAOYSA-L
copper ii sulfate pentahydrate, copper sulfate pentahydrate, cupric sulfate pentahydrate, blue vitriol, calcanthite, bluestone, copper 2+ sulfate pentahydrate, triangle, vencedor, blue copperas
24463
CHEBI:31440
copper;sulfate;pentahydrate
O.O.O.O.O.[Cu++].[O-]S([O-])(=O)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 110.0°C
Color Blue
Physical Form Crystalline Powder and/or Crystals
Quantity 1 kg
Assay Percent Range 97.5% min. (Iodometry)
Linear Formula CuSO4·5H2O
Fieser 01,164; 02,89; 05,162; 06,141; 14,267
Merck Index 15, 2643
Solubility Information Solubility in water: 320g/L (20°C). Other solubilities: soluble in methanol, slightly soluble in alcohol and glycerol, insoluble in ethanol
Formula Weight 249.68
Percent Purity 98%
Grade Extra Pure
Packaging Plastic Bottle
Chemical Name or Material Copper(II) sulfate pentahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Copper(II) sulfate pentahydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P362-Take off contaminated clothing.
  • P273-Avoid release to the environment.
Supplemental information
  • MIXTURE LIST-Contains : Copper (II) sulfatepentahydrate (1:1:5), Cupric sulfate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'