missing translation for 'onlineSavingsMsg'
Learn More

Copper(I) chloride, 97%, Thermo Scientific Chemicals

Catalog Number p-3256996
Click to view available options
Quantity:
500 g
2 kg
7758-89-6
ClCu
98.996
MFCD00010971
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]
7758-89-6
ClCu
98.996
MFCD00010971
OXBLHERUFWYNTN-UHFFFAOYSA-M
cuprous chloride, copper i chloride, copper chloride cucl, copper monochloride, dicopper dichloride, copper 1+ chloride, cucl, chlorid medny czech
62652
CHEBI:53472
chlorocopper
Cl[Cu]

It is precursor to many copper compounds including copper oxychloride and many organocuprate compounds of synthetic interest. It is used as a catalyst as well as a reagent in many organic reactions including Gatterman-Koch, Sandmeyer, Grignard and Gilman reactions. It catalyzes 1,4-addition of Grignard reagents to alpha,beta-unsaturated ketones. It is also used as a doping salt to enhance the cationization of polystyrene in matrix-assisted laser desorption/ionization mass spectrometry. It is also an intermediate formed in the Wacker process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 1490°C
Melting Point 430°C
Color Gray to Green
Physical Form Powder
Quantity 500 g
UN Number UN2802
Merck Index 14,2660
Solubility Information Very soluble in concentrated HCl. Soluble in ammonium hydroxide. Insoluble in ethanol and acetone. Sparingly soluble in water.
Formula Weight 98.99
Percent Purity 97%
Refractive Index 1.93
Sensitivity Air and moisture sensitive
Density 4.14
Chemical Name or Material Copper(I) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'