Learn More
Tenak™ Comfort Rack for Chest Freezers, 100 mm Box Height
Comfort Rack suitable for use with all chest freezer models.
77.80 € - 86.30 €
Specifications
Includes Spring Locks
Material Stainless Steel
For Use With (Equipment) Chest Frseezers
Height (Metric) 100 mm (Box)
View More Specs
Products 3
Product Code Brand Holds Price Quantity & Availability  
Product Code Brand Holds Price Quantity & Availability  
15200854
View Documents Promotion Details
Tenak™
TE24562
Boxes (4)
80.10 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-08-2024
to see stock.
Add to basket
15220854
View Documents Promotion Details
Tenak™
TE24564
Boxes (5)
86.30 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-08-2024
to see stock.
Add to basket
15280844
View Documents Promotion Details
Tenak™
TE24560
Boxes (3)
77.80 €
Each
View alternative products
Save Up to 
»
Estimated Shipment: 29-08-2024
to see stock.
Add to basket
Description
Description
  • Racks with spring locks mounded on each shelf to secure the cryoboxes
  • Can be used with most standard height cryoboxes in cardboard, polypropylene, and polycarbonate
  • Measure the inside height of your freezer when choosing the right rack
  • Constructed of stainless steel
Specifications
Specifications
Spring Locks
Stainless Steel
Chest Frseezers
100 mm (Box)