Learn More
COL6A6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560958
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: ALREVEHYFR PDMGSRINTG TPQVLLVLTD GQSQDEVAQA AEALRHRGID IYSVGIGDVD DQQLIQITGT AEKKLTVHNF DELKKVNKRI VRN Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 90%.
COL6A6 Polyclonal antibody specifically detects COL6A6 in Human samples. It is validated for Immunohistochemistry.Specifications
COL6A6 | |
Polyclonal | |
Unconjugated | |
COL6A6 | |
COL6A6; Collagen alpha-6(VI) chain; collagen type VI alpha 6; collagen, type VI, alpha 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
131873 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
A6NMZ7 | |
COL6A6 | |
Recombinant protein corresponding to Human COL6A6 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only