missing translation for 'onlineSavingsMsg'
Learn More

Coelenterazine, native, Aequorea sp., Thermo Scientific Chemicals

Catalog Number p-7076701
Click to view available options
Quantity:
1 mg
10 mg
55779-48-1
C26H21N3O3
423.472
MFCD00467176
YHIPILPTUVMWQT-UHFFFAOYSA-N
coelenterazine, oplophorus luciferin, unii-3o1cb88rrd, clzn, coelenterate luciferin, coelenterazine, native, 3o1cb88rrd, 8-benzyl-2-4-hydroxybenzyl-6-4-hydroxyphenyl imidazo 1,2-a pyrazin-3 7h-one, 2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazo 1,2-a pyrazin-3-7h-one, 3,2-dihydro-2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazolo 1,2-a pyrazin-3-one
2830
CHEBI:2311
8-benzyl-6-(4-hydroxyphenyl)-2-[(4-hydroxyphenyl)methyl]-7H-imidazo[1,2-a]pyrazin-3-one
C1=CC=C(C=C1)CC2=C3N=C(C(=O)N3C=C(N2)C4=CC=C(C=C4)O)CC5=CC=C(C=C5)O
55779-48-1
C26H21N3O3
423.472
MFCD00467176
YHIPILPTUVMWQT-UHFFFAOYSA-N
coelenterazine, oplophorus luciferin, unii-3o1cb88rrd, clzn, coelenterate luciferin, coelenterazine, native, 3o1cb88rrd, 8-benzyl-2-4-hydroxybenzyl-6-4-hydroxyphenyl imidazo 1,2-a pyrazin-3 7h-one, 2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazo 1,2-a pyrazin-3-7h-one, 3,2-dihydro-2-p-hydroxybenzyl-6-p-hydroxyphenyl-8-benzylimidazolo 1,2-a pyrazin-3-one
2830
CHEBI:2311
8-benzyl-6-(4-hydroxyphenyl)-2-[(4-hydroxyphenyl)methyl]-7H-imidazo[1,2-a]pyrazin-3-one
C1=CC=C(C=C1)CC2=C3N=C(C(=O)N3C=C(N2)C4=CC=C(C=C4)O)CC5=CC=C(C=C5)O

A sensitive and specific chemiluminescence probe of the superoxide anion

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Packaging Plastic bottle
Quantity 1 mg
Sensitivity Light and moisture sensitive
Solubility Information Soluble in methanol or ethanol
Formula Weight 423.46
Physical Form Solid
Chemical Name or Material Coelenterazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'