missing translation for 'onlineSavingsMsg'
Learn More

CLUH Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA565101

Additional Details : Weight : 0.01000kg

Product Code. 15927874

  • 497.00 € / 100µL
Estimated Shipment: 29-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: SRLGYEEHIP GQTRDWNEEL QTTRELPRKN LPERLLRERA IFKVHSDFTA AATRGAMAVI DGNVMAINPS EETKMQMFIW NNIFFSLGFD VRDHYKDFGG DVAAYVAPTN DLNGVRTYNA VDVEGLYTLG TVVVDYRGYR VTAQSIIPGI

CLUH Polyclonal antibody specifically detects CLUH in Human samples. It is validated for Immunocytochemistry.
Specifications

Specifications

CLUH
Polyclonal
Unconjugated
CLUH
1300001I01Rik; cb498; CG8443-like; CLU1; cluh; cluha; clustered mitochondria (cluA/CLU1) homolog; clustered mitochondria (cluA/CLU1) homolog a; clustered mitochondria homolog; clustered mitochondria protein homolog; KIAA0664; LOW QUALITY PROTEIN: clustered mitochondria protein homolog; mKIAA0664; RGD1307222; sb:cb498; Unknown (protein for IMAGE:6905374); wu:fb77e08; wu:fd36f07; wu:fe05g06; wu:fi27b10; zgc:152873
Rabbit
Antigen affinity chromatography
RUO
23277
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O75153
CLUH
Recombinant Human CLUH
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

For Research Use Only