missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ cis-Stilbeneboronic acid pinacol ester, 99%
Click to view available options
Quantity:
250 mg
1 g
Specifications
Melting Point | 92°C to 94°C |
Quantity | 250 mg |
Beilstein | 9269561 |
Formula Weight | 306.22 |
Percent Purity | 99% |
Chemical Name or Material | cis-Stilbeneboronic acid pinacol ester |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'