missing translation for 'onlineSavingsMsg'
Learn More

cis-6-(Benzyloxycarbonylamino)-3-cyclohexene-1-carboxylic acid, 97%, Thermo Scientific™

Catalog Number 15427686
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
124753-65-7
C15H16NO4
274.30
MFCD01863246,MFCD01863246,MFCD03844603
QIAAZWPFGHTXNZ-OLZOCXBDSA-M
1s,6r-6-benzyloxy carbonyl amino cyclohex-3-ene-1-carboxylic acid, cis-6-benzyloxycarbonylaminocyclohex-3-enecarboxylic acid, cis-6-benzyloxy carbonyl amino cyclohex-3-enecarboxylic acid, 1s-6beta-benzyloxycarbonylamino-3-cyclohexene-1beta-carboxylic acid, 3-cyclohexene-1-carboxylic acid, 6-phenylmethoxy carbonyl amino-, 1r,6s-rel-9ci
21356434
(1S,6R)-6-(phenylmethoxycarbonylamino)cyclohex-3-ene-1-carboxylic acid
[O-]C(=O)[C@@H]1CC=CC[C@@H]1NC(=O)OCC1=CC=CC=C1
This item is not returnable. View return policy
124753-65-7
C15H16NO4
274.30
MFCD01863246,MFCD01863246,MFCD03844603
QIAAZWPFGHTXNZ-OLZOCXBDSA-M
1s,6r-6-benzyloxy carbonyl amino cyclohex-3-ene-1-carboxylic acid, cis-6-benzyloxycarbonylaminocyclohex-3-enecarboxylic acid, cis-6-benzyloxy carbonyl amino cyclohex-3-enecarboxylic acid, 1s-6beta-benzyloxycarbonylamino-3-cyclohexene-1beta-carboxylic acid, 3-cyclohexene-1-carboxylic acid, 6-phenylmethoxy carbonyl amino-, 1r,6s-rel-9ci
21356434
(1S,6R)-6-(phenylmethoxycarbonylamino)cyclohex-3-ene-1-carboxylic acid
[O-]C(=O)[C@@H]1CC=CC[C@@H]1NC(=O)OCC1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Formula Weight 275.29
Percent Purity 97%
Quantity 1 g
Physical Form Solid
Chemical Name or Material cis-6-(Benzyloxycarbonylamino)-3-cyclohexene-1-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'