missing translation for 'onlineSavingsMsg'
Learn More

cis-3-Acetyl-2,2-dimethylcyclobutaneacetic acid, 97%, Thermo Scientific™

Catalog Number 15427476
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
64396-97-0
C10H16O3
184.24
MFCD00001327,MFCD09864988
SIZDUQQDBXJXLQ-UHFFFAOYNA-N
cis-dl-pinonic acid, 1s,3s-3-acetyl-2,2-dimethylcyclobutyl acetic acid, pinonic acid, cis dl, 2-1s,3s-3-acetyl-2,2-dimethylcyclobutyl-acetic acid, 2-1s,3s-3-acetyl-2,2-dimethylcyclobutyl acetic acid, cis-3-acetyl-2,2-dimethylcyclobutaneacetic acid, 1s,3s-2,2-dimethyl-3-acetylcyclobutyl acetic acid, 1s,3s-+-cis-3-acetyl-2,2-dimethylcyclobutaneacetic acid
239867
2-[(1S,3S)-3-acetyl-2,2-dimethylcyclobutyl]acetic acid
CC(=O)C1CC(CC(O)=O)C1(C)C
This item is not returnable. View return policy
64396-97-0
C10H16O3
184.24
MFCD00001327,MFCD09864988
SIZDUQQDBXJXLQ-UHFFFAOYNA-N
cis-dl-pinonic acid, 1s,3s-3-acetyl-2,2-dimethylcyclobutyl acetic acid, pinonic acid, cis dl, 2-1s,3s-3-acetyl-2,2-dimethylcyclobutyl-acetic acid, 2-1s,3s-3-acetyl-2,2-dimethylcyclobutyl acetic acid, cis-3-acetyl-2,2-dimethylcyclobutaneacetic acid, 1s,3s-2,2-dimethyl-3-acetylcyclobutyl acetic acid, 1s,3s-+-cis-3-acetyl-2,2-dimethylcyclobutaneacetic acid
239867
2-[(1S,3S)-3-acetyl-2,2-dimethylcyclobutyl]acetic acid
CC(=O)C1CC(CC(O)=O)C1(C)C

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 104°C to 107°C
Quantity 250 mg
Formula Weight 184.23
Percent Purity 97%
Physical Form Solid
Chemical Name or Material cis-3-Acetyl-2,2-dimethylcyclobutaneacetic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'