missing translation for 'onlineSavingsMsg'
Learn More

Chrysin, 99+%, Thermo Scientific Chemicals

Catalog Number p-3757308
35.00 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
480-40-0
C15H10O4
254.24
MFCD00006834
RTIXKCRFFJGDFG-UHFFFAOYSA-N
chrysin, 5,7-dihydroxyflavone, chrysine, 5,7-dihydroxy-2-phenyl-4h-chromen-4-one, crysin, 4h-1-benzopyran-4-one, 5,7-dihydroxy-2-phenyl, unii-3cn01f5zj5, flavone, 5,7-dihydroxy, 5,7-dihydroxy-2-phenyl-4h-1-benzopyran-4-one, chembl117
5281607
CHEBI:75095
5,7-dihydroxy-2-phenylchromen-4-one
C1=CC=C(C=C1)C2=CC(=O)C3=C(C=C(C=C3O2)O)O
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
480-40-0
C15H10O4
254.24
MFCD00006834
RTIXKCRFFJGDFG-UHFFFAOYSA-N
chrysin, 5,7-dihydroxyflavone, chrysine, 5,7-dihydroxy-2-phenyl-4h-chromen-4-one, crysin, 4h-1-benzopyran-4-one, 5,7-dihydroxy-2-phenyl, unii-3cn01f5zj5, flavone, 5,7-dihydroxy, 5,7-dihydroxy-2-phenyl-4h-1-benzopyran-4-one, chembl117
5281607
CHEBI:75095
5,7-dihydroxy-2-phenylchromen-4-one
C1=CC=C(C=C1)C2=CC(=O)C3=C(C=C(C=C3O2)O)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 284.0°C to 290.0°C
Color Yellow
Loss on Drying 1.0% max.
Infrared Spectrum Authentic
Assay Percent Range 99% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Beilstein 18,124
Merck Index 15,226
Formula Weight 254.24
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Chrysin
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'