missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ CGA (Human) Recombinant Protein
Human CGA full-length ORF ( AAH10957, 1 a.a. - 116 a.a.) recombinant protein with GST-tag at N-terminal.
Marke: Abnova™ H00001081-P01.10ug
Weitere Details : Gewicht : 0.00010kg
Beschreibung
- Sequence: MDYYRKYAAIFLVTLSVFLHVLHSAPDVQDCPECTLQENPLFSQPGAPILQCMGCCFSRAYPTPLRSKKTMLVQKNVTSESTCCVAKSYNRVTVMGGFKVENHTACHCSTCYYHKS
Spezifikation
AAH10957 | |
glycoprotein hormones, alpha polypeptide | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CGA | |
GST |
1081 | |
Wheat germ expression system | |
10 μg | |
CG-ALPHA, FSHA, GPHA1, GPHa, HCG, LHA, TSHA | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |